MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Selective transport of Li+ across lipid bilayer membranes mediated by an ionophore of novel design (ETH1644).
Selective transport of Li+ across lipid bilayer membranes mediated by an ionophore of novel design (ETH1644).
The Journal of membrane biology (1985-01-01)
A Zeevi, R Margalit
ABSTRACT
The neutral noncyclic, lithium-selective ionophore ETH1644, which is structurally different from previously available ionophores of this type, is a selective carrier of Li+ in lipid bilayer membranes of various lipid composition. The ionophore forms a 2:1 carrier/cation complex, and the rate-limiting step in the overall transport process is the diffusion of the carrier/ion complex across the membrane. The selectivity sequence for lithium vs. other ions normally found in biological systems is: Li+ (1) greater than Na+ (0.017) greater than or equal to K+ (0.017) greater than Cl- (0.001), Ca2+ and Mg2+ are impermeant. At neutral pH protons do not interfere with the Li+-carrying ability of this ionophore. On the basis of structural differences and supported by conductance data, it is argued that the improved selectivity of Li+ over the other alkali cations is due more to a decrease in the affinities of the ionophore for the latter cations that to an increase of its affinity to Li+. This ionophore can also act as a carrier of biogenic amines (catecholes, indoles and derivatives), with the structure of the permeant species and mechanism of permeation similar to that observed with the alkali cations. The selectivity sequence is: tryptamine (18.1) greater than phenylethylamine (11.6) greater than tyramine (2.4) greater than Li+ (1) greater than serotonin (0.34) greater than epinephrine (0.09) greater than dopamine (0.05) greater than norepinephrine (0.02), showing the ionophore to be more selective to Li+ than to any of the neurotransmitters studied.