MilliporeSigma
  • [Effect of oxygen on the irreversible radiochemical transformations of protoporphyrin in 2n. H2SO4].
[Effect of oxygen on the irreversible radiochemical transformations of protoporphyrin in 2n. H2SO4].
Radiobiologiia (1986-09-01)
I I Makarenkova
ABSTRACT
The mechanism of the inverse oxygen effect in the irreversible radiation-induced chemical alterations in protoporphyrin in sulfuric acid solution was investigated. The decrease of the molecular decay yield was shown to associate with the ability of oxygen to exclude H atoms from the radiation reactions and to inhibit the development of the protoporphyrin transformation chain process.
Sigma-Aldrich
Protoporphyrin IX dimethyl ester, ~90% (HPLC)