MilliporeSigma
  • Desaminopenicillamine tocinoic acid derivatives--inhibitors of oxytocin.
Desaminopenicillamine tocinoic acid derivatives--inhibitors of oxytocin.
International journal of peptide and protein research (1986-08-01)
U Roy, D Gazis, J D Glass, I L Schwartz
ABSTRACT
Tocinoic acid analogs with penicillamine in place of one or both of the cysteine residues have been studied and [1-beta-mercaptopropionic acid, 6-penicillamine] tocinoic acid (dPen6TA) and [1-beta,beta-dimethyl-beta-mercaptopropionic acid, 6-penicillamine] tocinoic acid (dPen1Pen6TA) have been synthesized in solution. Biological activities of these 2 compounds and those of the previously synthesized [1-beta,beta-dimethyl-beta-mercaptopropionic acid] tocinoic acid (dPen1TA) have been assayed. It was found that dPen1TA and dPen1Pen6TA, both of which have a beta,beta-dimethyl-beta-mercaptopropionic acid in position 1, are strong inhibitors of the uterine activity of oxytocin in vitro (without Mg2+) with pA2 values of 7.1 and 7.8, respectively, whereas dPen6TA with penicillamine in position 6 is a mild agonist.