MilliporeSigma
  • Sites of methylation of DNA bases by the action of organophosphorus insecticides in vitro.
Sites of methylation of DNA bases by the action of organophosphorus insecticides in vitro.
Acta biochimica Polonica (1986-01-01)
R Wiaderkiewicz, Z Walter, W Reimschussel
ABSTRACT
Methylation in vitro of DNA by three methyl-14C-labelled organophosphorus insecticides has been studied. The ability of methylbromphenvinphos, methylparathion and malathion to methylate N-7 of guanine in DNA can be expressed as 100:40:15. Among the methylation products, no O6-methylguanine, a known mutagen, was found. Both in the reaction with dsDNA and with ssDNA 7-methyl-guanine was the main methylation product. However, all methyl derivatives of adenine (3-methyladenine, 1-methyladenine and 7-methyladenine) constituted about 40% and 50% of all methylation products in the case of dsDNA and ssDNA, respectively. The only methyl derivative of pyrimidine we have identified was 3-methylcytosine. In the case of dsDNA 3-methylcytosine appeared in small amounts but in the alkylated ssDNA 3-methylcytosine C constituted about 20% of all alkylation products.