MilliporeSigma
  • A method for the continuous measurement of respiration and vapor uptake in rats.
A method for the continuous measurement of respiration and vapor uptake in rats.
American Industrial Hygiene Association journal (1987-06-01)
A R Dahl, T P Gugliotta, R L Hanson, J L Mauderly, S J Rothenberg
ABSTRACT
A method for determining the uptake of inhaled vapors in rats is described. The method uses gas chromatographic data to enable calculations of vapor concentration differences upstream and downstream from a rat exposed to the vapor by the nose-only route. Respiratory data are recorded simultaneously. The data can be used to determine the rate of uptake, uptake as a fraction of vapor inhaled and the effects of vapor inhalation on respiration.
Sigma-Aldrich
2,3,4-Trimethylpentane, 98%