MilliporeSigma
  • Correlation of fluorescent tracer measurements of dermal exposure and urinary metabolite excretion during occupational exposure to malathion.
Correlation of fluorescent tracer measurements of dermal exposure and urinary metabolite excretion during occupational exposure to malathion.
American Industrial Hygiene Association journal (1988-09-01)
R A Fenske
ABSTRACT
Nineteen workers conducting mixing and high-volume airblast applications of the organophosphorus pesticide malathion were monitored simultaneously by biological monitoring and fluorescent tracer evaluation of dermal exposure. Complete 72-hr urine samples were collected and analyzed for dimethylthiophosphate and dimethyldithiophosphate metabolites. Dermal exposure was measured through the addition of a fluorescent tracer to the tank mix, subsequent examination of the skin surface under long-wave ultraviolet light, and fluorescence quantification with a video imaging system. Dermal exposure to applicators was correlated highly with total metabolite excretion (r = 0.91). Mixer exposure was not correlated significantly (r = 0.73) because of wide scatter in the data and the small number of workers monitored. Applicator exposures were more than 3 times higher than mixer exposures, reflecting the high exposure potential inherent in airblast spraying. Exposure to regions protected by gloves or clothing was more than 75% of total exposure for both mixers and applicators. These results provide evidence that the fluorescent tracer technique is a valid methodology for measuring relative levels of dermal exposure during agricultural work activities. The technique also holds promise as a quantitative procedure for evaluating the effectiveness of engineering control strategies and protective clothing performance.