MilliporeSigma
  • Effect of the suicide substrate 3,5-diethoxycarbonyl-2,6-dimethyl-4-ethyl-1,4-dihydropyridine on the metabolism of xenobiotics and on cytochrome P-450 apoproteins.
Effect of the suicide substrate 3,5-diethoxycarbonyl-2,6-dimethyl-4-ethyl-1,4-dihydropyridine on the metabolism of xenobiotics and on cytochrome P-450 apoproteins.
Molecular pharmacology (1986-01-01)
T R Tephly, K A Black, M D Green, B L Coffman, G A Dannan, F P Guengerich
ABSTRACT
Treatment of rats with the cytochrome P-450 suicide substrate, 3,5-diethoxycarbonyl-2,6-dimethyl-4-ethyl-1,4-dihydropyridine (DDEP), produced a 95% inhibition of the in vivo demethylation of either aminopyrine or morphine within 2 hr. One-carbon metabolism of formaldehyde or formate to carbon dioxide was not altered. DDEP also produced a time-dependent decrease in total hepatic microsomal cytochrome P-450 but had no effect on either NADPH-cytochrome c reductase or p-nitrophenol glucuronyl-transferase activities up to 24 hr after administration. A rapid decrease in rat liver microsomal aniline hydroxylation and ethoxyresorufin deethylation was observed in vitro following DDEP administration. Although in vitro testosterone metabolism to 16 alpha-, 16 beta-, and 2 alpha-hydroxy metabolites was depressed profoundly by DDEP in microsomes from untreated and 3-methylcholanthrene-treated animals, 7 alpha-hydroxylation of testosterone was much less affected. Immunochemical quantification of various microsomal cytochrome P-450 protein moieties showed that cytochromes P-450 beta NF-B, P-450UT-A, P-450PCN-E, and P-450PB-C were decreased in hepatic microsomes from DDEP-treated rats. However, the protein moiety of cytochrome P-450UT-H was not diminished and the immunoreactive protein for cytochromes P-450UT-F, P-450PB-B, and P-450ISF-G was only slightly decreased. These results show that DDEP treatment leads to marked decreases in holoprotein and apoproteins of many but not all hepatic microsomal cytochrome P-450 isozymes.