MilliporeSigma
  • 5-Halogeno-3'-fluoro-2',3'-dideoxyuridines as inhibitors of human immunodeficiency virus (HIV): potent and selective anti-HIV activity of 3'-fluoro-2',3'-dideoxy-5-chlorouridine.
5-Halogeno-3'-fluoro-2',3'-dideoxyuridines as inhibitors of human immunodeficiency virus (HIV): potent and selective anti-HIV activity of 3'-fluoro-2',3'-dideoxy-5-chlorouridine.
Molecular pharmacology (1989-05-01)
J Balzarini, A Van Aerschot, R Pauwels, M Baba, D Schols, P Herdewijn, E De Clercq
ABSTRACT
The novel 5-chloro-, 5-bromo-, and 5-iodo-derivatives of 3'-fluoro-2',3'-dideoxyuridine (FddUrd), designated FddCIUrd, FddBrUrd, and FddIUrd, respectively, have been synthesized and evaluated for their antiretrovirus activity against human immunodeficiency virus (HIV) and murine Moloney sarcoma virus. All three 5-halogeno-FddUrd analogues inhibited HIV-1 replication in MT4 cells with an effective dose (ED50) of about 0.2-0.4 microM. However, FddCIUrd was markedly more selective in its anti-HIV-1 activity than FddBrUrd or FddIUrd. The selectivity index of FddCIUrd was similar to that of 3'-azido-2',3'-dideoxythymidine (AZT) when evaluated in parallel (1408 and 1603, respectively). The FddUrd derivatives also had a marked inhibitory effect on HIV-2 replication in MT4 cells and HIV-1 induced antigen expression in HUT-78 cells. However, neither FddUrd nor its 5-halogeno derivatives were inhibitory to Moloney sarcoma virus-induced transformation of murine C3H cells. The anti-HIV-1 activity of FddUrd, FddCIUrd, FddBrUrd, and FddIUrd was reversed by the addition of thymidine and 2'-deoxycytidine. The 5-halogeno-FddUrd analogues had a markedly higher affinity for MT4 thymidine kinase than FddUrd (Ki/Km, 4.0-4.7, as compared with 302 for FddUrd).