Sigma-Aldrich
Platinum black, black, powder, ≤20 μm, ≥99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.127 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, gauze, 100 mesh, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum black, fuel cell grade, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, shot, ≤3 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.127 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.076 mm, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 2.0 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.25 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.1 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Oxaliplatin, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, powder, 99.995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, powder (coarse), 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 1.0 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.25 mm, thermocouple grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.025 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 1.5 mm, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.05 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, foil, thickness 0.125-0.135 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, sponge, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.20 mm, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.10 mm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, wire, diam. 0.5 mm, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Platinum, gauze, 52 mesh, 99.9% trace metals basis