MilliporeSigma
  • Association of a functional FAAH polymorphism with methamphetamine-induced symptoms and dependence in a Malaysian population.
Association of a functional FAAH polymorphism with methamphetamine-induced symptoms and dependence in a Malaysian population.
Pharmacogenomics (2013-04-06)
Maw Shin Sim, Ahmad Hatim, Gavin Paul Reynolds, Zahurin Mohamed
ABSTRACT
FAAH is a membrane enzyme that terminates the activity of a large class of endogenous signaling lipids. Recent studies suggest that the FAAH Pro129Thr polymorphism is a common mutation in the FAAH gene that is significantly associated with drug-addictive traits. This study investigated the association of the Pro129Thr polymorphism of the FAAH gene with methamphetamine dependence, methamphetamine-induced psychosis, manic episodes and panic disorder in a Malaysian population. This polymorphism was genotyped in 232 male methamphetamine-dependent subjects and in 241 male controls of four different ethnicities: Malay, Chinese, Kadazan-Dusun and Bajau. Intergroup statistical analyses were performed by using the χ(2)-square test and Fisher's exact test, where necessary. In cases of multiple comparisons, the Bonferroni correction was performed. Our results indicated that the FAAH Pro129Thr polymorphism showed a significant association with risk of methamphetamine dependence in the pooled subjects (odds ratio [OR]: 2.017; p < 0.001) and in the Malay (OR: 2.829; p < 0.001) and Chinese (OR: 3.685; p < 0.001) groups. We also found an association of this polymorphism with episodes of methamphetamine-induced mania in the Malay group (OR: 2.836; p = 0.035). However, there was no association between this polymorphism and age of onset of drug use or the occurrence of methamphetamine-induced psychosis or of panic disorder. Our findings suggest that the FAAH Pro129Thr polymorphism may contribute to methamphetamine dependence in the Malay and Chinese ethnic groups.
Product Description
Supelco
S(+)-Methamphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®