MilliporeSigma
  • A non-destructive genotyping system from a single seed for marker-assisted selection in watermelon.
A non-destructive genotyping system from a single seed for marker-assisted selection in watermelon.
Genetics and molecular research : GMR (2013-04-03)
G Meru, D McDowell, V Waters, A Seibel, J Davis, C McGregor
ABSTRACT
Genomic tools for watermelon breeding are becoming increasingly available. A high throughput genotyping system would facilitate the use of DNA markers in marker-assisted selection. DNA extraction from leaf material requires prior seed germination and is often time-consuming and cost prohibitive. In an effort to develop a more efficient system, watermelon seeds of several genotypes and various seed sizes were sampled by removing ⅓ or ½ sections from the distal ends for DNA extraction, while germinating the remaining proximal parts of the seed. Removing ⅓ of the seed from the distal end had no effect on seed germination percentage or seedling vigor. Different DNA extraction protocols were tested to identify a method that could yield DNA of sufficient quality for amplification by polymerase chain reaction. A sodium dodecyl sulfate extraction protocol with 1% polyvinylpyrrolidone yielded DNA that could be amplified with microsatellite primers and was free of pericarp contamination. In this study, an efficient, non-destructive genotyping protocol for watermelon seed was developed.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Kollidon® 25
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Polyvinylpyrrolidone, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture