MilliporeSigma
  • Dual behavior of isatin-based cyclic ketimines with dicarbomethoxy carbene: expedient synthesis of highly functionalized spirooxindolyl oxazolidines and pyrrolines.
Dual behavior of isatin-based cyclic ketimines with dicarbomethoxy carbene: expedient synthesis of highly functionalized spirooxindolyl oxazolidines and pyrrolines.
Organic letters (2013-03-13)
T Rajasekaran, G Karthik, B Sridhar, B V Subba Reddy
ABSTRACT
A highly stereo-, regio-, and chemoselective method has been devised for the synthesis of a wide range of spirooxindolyl oxazolidines via an intermolecular 1,3-dipolar cycloaddition of carbonyl ylides generated from dimethyl diazomalonate and aromatic aldehydes, with cyclic ketimines using 5 mol % of Rh2(OAc)4 under mild conditions. Similarly, highly functionalized spirooxindolyl pyrrolines have also been prepared through 1,3-dipolar cycloaddition of azomethine ylides generated from dimethyl diazomalonate and cyclic ketimines, with dimethyl acetylenedicarboxylate.
Sigma-Aldrich
Isatin, 97%