MilliporeSigma
  • Fungal metabolites of xanthohumol with potent antiproliferative activity on human cancer cell lines in vitro.
Fungal metabolites of xanthohumol with potent antiproliferative activity on human cancer cell lines in vitro.
Bioorganic & medicinal chemistry (2013-02-26)
Tomasz Tronina, Agnieszka Bartmańska, Beata Filip-Psurska, Joanna Wietrzyk, Jarosław Popłoński, Ewa Huszcza
ABSTRACT
Xanthohumol (1) and xanthohumol D (2) were isolated from spent hops. Isoxanthohumol (3) was obtained from xanthohumol by isomerisation in alkaline solution. Six metabolites were obtained as a result of transformation of xanthohumol (1) by selected fungal cultures. Their structures were established on the basis of their spectral data. One of them: 2″-(2'''-hydroxyisopropyl)-dihydrofurano-[4″,5″:3',4']-4',2-dihydroxy-6'-methoxy-α,β-dihydrochalcone (6) has not been previously reported in the literature. The antioxidant properties of hops flavonoids and xanthohumol derivatives were investigated using the 2,2'-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging method. The effects of these compounds on proliferation of MCF-7, PC-3 and HT-29 human cancer cell lines were determined by the SRB assay. With the exception of one metabolite, all tested compounds showed antiproliferative activity against the tested human cancer lines. α,β-Dihydroxanthohumol (4), obtained through the biotransformation of xanthohumol, showed higher antiproliferative activity against MCF-7 human breast carcinoma cell line than cisplatin, a widely used anticancer therapeutic agent, and a comparably high activity against PC-3 human prostate cancer cell line.
Product Description
Xanthohumol, primary reference standard
Pricing and availability is not currently available.