MilliporeSigma
[Vertigo: patient management tactics].
Likars'ka sprava (2013-01-29)
I S Vasyl'chuk, O O Kozlova, K F Trinus, Iu I Holovchenko, T M Slobodin
ABSTRACT
To avoid serious causes of dizziness and establishing preliminary diagnosis is usually enough sighting history taking and physical examination of the patient. Great importance is self-presenting patients with complaints of dizziness. In recent years, for treatment of various types vestybulopatiy, but also for the prevention of attacks, successfully used betahistyn hydrochloride, which improves the quality of care and well-being of the patient.