MilliporeSigma
Temperature dependent optical properties of PbS nanocrystals.
Nanotechnology (2012-06-19)
M N Nordin, Juerong Li, S K Clowes, R J Curry
ABSTRACT
A comprehensive study of the optical properties of PbS nanocrystals (NCs) is reported that includes the temperature dependent absorption, photoluminescence (PL) and PL lifetime in the range of 3-300 K. The absorption and PL are found to display different temperature dependent behaviour though both redshift as temperature is reduced. This results in a temperature dependent Stokes shift which increases from ∼75 meV at 300 K with reducing temperature until saturating at ∼130 meV below ∼150 K prior to a small reduction to 125 meV upon cooling from 25 to 3 K. The PL lifetime is found to be single exponential at 3 K with a lifetime of τ(1) = 6.5 μs. Above 3 K biexponential behaviour is observed with the lifetime for each process displaying a different temperature dependence. The Stokes shift is modelled using a three-level rate equation model incorporating temperature dependent parameter values obtained via fitting phenomenological relationships to the observed absorption and PL behaviour. This results in a predicted energy difference between the two emitting states of ∼6 meV which is close to the excitonic exchange energy splitting predicted theoretically for these systems.
Sigma-Aldrich
Lead(II) selenide, 99.99%