MilliporeSigma
  • Thrombotic stroke in the anesthetized monkey (Macaca mulatta): characterization by MRI--a pilot study.
Thrombotic stroke in the anesthetized monkey (Macaca mulatta): characterization by MRI--a pilot study.
Cerebrovascular diseases (Basel, Switzerland) (2012-02-22)
Maxime Gauberti, Pauline Obiang, Pierre Guedin, Anne Balossier, Clément Gakuba, Anne Sophie Diependaele, Laurent Chazalviel, Denis Vivien, Alan R Young, Véronique Agin, Cyrille Orset
ABSTRACT
The lack of a relevant stroke model in large nonhuman primates hinders the development of innovative diagnostic/therapeutic approaches concerned with this cerebrovascular disease. Our objective was to develop a novel and clinically relevant model of embolic stroke in the anesthetized monkey that incorporates readily available clinical imaging techniques and that would allow the possibility of drug delivery including strategies of reperfusion. Thrombin was injected into the lumen of the middle cerebral artery (MCA) in 12 anesthetized (sevoflurane) male rhesus macaques (Macaca mulatta). Sequential MRI studies (including angiography, FLAIR, PWI, DWI, and gadolinium-enhanced T1W imaging) were performed in a 3T clinical MRI. Physiological and biochemical parameters were monitored throughout the investigations. Once standardized, the surgical procedure induced transient occlusion of the middle cerebral artery in all operated animals. All animals studied showed spontaneous reperfusion, which occurred some time between 2 h and 7 days post-ictus. Eighty percent of the studied animals showed diffusion/perfusion mismatch. The ischemic lesions at 24 h spared both superficial and profound territories of the MCA. Some animals presented hemorrhagic transformation at 7 days post-ictus. In this study, we developed a pre-clinically relevant model of embolic stroke in the anesthetized nonhuman primate.
Product Description
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard