MilliporeSigma
  • The role of testicular hormones and luteinizing hormone in spatial memory in adult male rats.
The role of testicular hormones and luteinizing hormone in spatial memory in adult male rats.
Hormones and behavior (2012-01-24)
Sarah E A McConnell, Juliet Alla, Elizabeth Wheat, Russell D Romeo, Bruce McEwen, Janice E Thornton
ABSTRACT
Attempts to determine the influence of testicular hormones on learning and memory in males have yielded contradictory results. The present studies examined whether testicular hormones are important for maximal levels of spatial memory in young adult male rats. To minimize any effect of stress, we used the Object Location Task which is a spatial working memory task that does not involve food or water deprivation or aversive stimuli for motivation. In Experiment 1 sham gonadectomized male rats demonstrated robust spatial memory, but gonadectomized males showed diminished spatial memory. In Experiment 2 subcutaneous testosterone (T) capsules restored spatial memory performance in gonadectomized male rats, while rats with blank capsules demonstrated compromised spatial memory. In Experiment 3, gonadectomized male rats implanted with blank capsules again showed compromised spatial memory, while those with T, dihydrotestosterone (DHT), or estradiol (E) capsules demonstrated robust spatial memory, indicating that T's effects may be mediated by its conversion to E or to DHT. Gonadectomized male rats injected with Antide, a gonadotropin-releasing hormone receptor antagonist which lowers luteinizing hormone levels, also demonstrated spatial memory, comparable to that shown by T-, E-, or DHT-treated males. These data indicate that testicular androgens are important for maximal levels of spatial working memory in male rats, that testosterone may be converted to E and/or DHT to exert its effects, and that some of the effects of these steroid hormones may occur via negative feedback effects on LH.