MilliporeSigma
  • Synthesis of multifunctional nanostructured zinc-iron mixed oxide photocatalyst by a simple solution-combustion technique.
Synthesis of multifunctional nanostructured zinc-iron mixed oxide photocatalyst by a simple solution-combustion technique.
ACS applied materials & interfaces (2011-12-29)
Gajendra Kumar Pradhan, Satyabadi Martha, K M Parida
ABSTRACT
A series of nanostructure zinc-iron mixed oxide photocatalysts have been fabricated by solution-combustion method using urea as the fuel, and nitrate salts of both iron and zinc as the metal source. Different characterization tools, such as X-ray diffraction (XRD), X-ray photoelectron spectroscopy (XPS), diffuse reflectance UV-visible spectra (DRUV-vis), electron microscopy, and photoelectrochemical measurement were employed to establish the structural, electronic, and optical properties of the material. Electron microscopy confirmed the nanostructure of the photocatalyst. The synthesized photocatalysts were examined towards photodegradation of 4-chloro-2-nitro phenol (CNP), rhodamine 6G (R6G), and photocatalytic hydrogen production under visible light (λ ≥ 400 nm). The photocatalyst having zinc to iron ratio of 50:50 showed best photocatalytic activity among all the synthesized photocatalysts.
Sigma-Aldrich
4-Chloro-2-nitrophenol, ≥97.0%
Pricing and availability is not currently available.