MilliporeSigma
Direct cupration of fluoroform.
Journal of the American Chemical Society (2011-12-06)
Alessandro Zanardi, Maxim A Novikov, Eddy Martin, Jordi Benet-Buchholz, Vladimir V Grushin
ABSTRACT
We have found the first reaction of direct cupration of fluoroform, the most attractive CF(3) source for the introduction of the trifluoromethyl group into organic molecules. Treatment of CuX (X = Cl, Br, I) with 2 equiv of MOR (M = K, Na) in DMF or NMP produces novel alkoxycuprates that readily react with CF(3)H at room temperature and atmospheric pressure to give CuCF(3) derivatives. The CuCl and t-BuOK (1:2) combination provides best results, furnishing the CuCF(3) product within seconds in nearly quantitative yield. As demonstrated, neither CF(3)(-) nor CF(2) mediate the Cu-CF(3) bond formation, which accounts for its remarkably high selectivity. The fluoroform-derived CuCF(3) solutions can be efficiently stabilized with TREAT HF to produce CuCF(3) reagents that readily trifluoromethylate organic and inorganic electrophiles in the absence of additional ligands such as phenanthroline. A series of novel Cu(I) complexes have been structurally characterized, including K(DMF)[Cu(OBu-t)(2)] (1), Na(DMF)(2)[Cu(OBu-t)(2)] (2), [K(8)Cu(6)(OBu-t)(12)(DMF)(8)(I)](+) I(-) (3), and [Cu(4)(CF(3))(2)(C(OBu-t)(2))(2)(μ(3)-OBu-t)(2)] (7).