MilliporeSigma
  • High-throughput screening of drug-lipid membrane interactions via counter-propagating second harmonic generation imaging.
High-throughput screening of drug-lipid membrane interactions via counter-propagating second harmonic generation imaging.
Analytical chemistry (2011-06-24)
Trang T Nguyen, John C Conboy
ABSTRACT
Here we report the use of counter-propagating second harmonic generation (SHG) to image the interactions between the local anesthetic tetracaine and a multicomponent planar supported lipid bilayer array in a label-free manner. The lipid bilayer arrays, prepared using a 3D continuous flow microspotter, allow the effects of lipid phase and cholesterol content on tetracaine binding to be examined simultaneously. SHG images show that tetracaine has a higher binding affinity to liquid-crystalline phase lipids than to solid-gel phase lipids. The presence of 28 mol % cholesterol decreased the binding affinity of tetracaine to bilayers composed of the mixed chain lipid, 1-steroyl-2-oleoyl-sn-glycero-3-phophocholine (SOPC), and the saturated lipids 1,2-dimyristoyl-sn-glycero-3-phophocholine (DMPC) and 1,2-dipamitoyl-sn-glycero-3-phophocholine (DPPC) while having no effect on diunsaturated 1,2-dioleoyl-sn-glycero-3-phophocholine (DOPC). The maximum surface excess of tetracaine increases with the degree of unsaturation of the phospholipids and decreases with cholesterol in the lipid bilayers. The paper demonstrates that SHG imaging is a sensitive technique that can directly image and quantitatively measure the association of a drug to a multicomponent lipid bilayer array, providing a high-throughput means to assess drug-membrane interactions.