MilliporeSigma
[Alkaloids from Senecio scandens].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-12-24)
Daopeng Tan, Ying Chen, Lili Ji, Guixin Chou, Zhengtao Wang
ABSTRACT
To investigate the alkaloids from Senecio scandens. Compounds were isolated with silica gel and Sephadex LH-20 column chromatography and their structures were determined by spectral analysis and chemical evidence. The hepatic cytotoxicity of isolated compounds was tested by MTT method in vitro. Six alkaloids were obtained and identified as adonifoline (1), 7-angeloylturneforcidine (2), hordenine (3), 1, 3, 6, 6-tetramethyl-5, 6, 7, 8-tetrahydro-isoquinolin-8-one (4), 4-(pyrrolidin-2-one) -phenyl acetic acid (5), (4-pyrrolidinophenyl) acetic acid (6). Compound 6 is a new natural product, compounds 3, 4 were obtained from the genus Senecio for the first time, compounds 2, 5 were obtained from this plant for the first time. Compound 1 showed significant growth inhibitory effect against hepatocyte at 100 micromol x L(-1).
Sigma-Aldrich
Hordenine, ≥97.0% (HPLC)