MilliporeSigma
  • Prenatal diagnosis of Morquio disease type A using a simple fluorometric enzyme assay.
Prenatal diagnosis of Morquio disease type A using a simple fluorometric enzyme assay.
Prenatal diagnosis (1990-02-01)
H Zhao, O P Van Diggelen, R Thoomes, J Huijmans, E Young, T Mazurczak, W J Kleijer
ABSTRACT
A new fluorogenic substrate, 4 methylumbelliferyl beta-D-6-sulphogalactoside, was used for the assay of galactose-6-sulphate sulphatase activity in chorionic villi, cultured villus cells, and amniocytes. The fluorometric assay is much more convenient than the conventional assay using radiolabelled, sulphated oligosaccharides. Both types of substrate were used in the prenatal diagnosis of three pregnancies at risk for Morquio type A disease using amniocytes. These enzyme tests, as well as electrophoresis of glycosaminoglycans in the amniotic fluid, indicated affected fetuses in two pregnancies and a non-affected fetus in one.
Sigma-Aldrich
4-Methylumbelliferyl β-D-galactopyranoside-6-sulfate sodium salt, ≥90% (HPLC)