MilliporeSigma
  • Formation of pyrazine derivatives from D-glucosamine and their deoxyribonucleic acid (DNA) strand breakage activity.
Formation of pyrazine derivatives from D-glucosamine and their deoxyribonucleic acid (DNA) strand breakage activity.
Chemical & pharmaceutical bulletin (1991-03-01)
K Sumoto, M Irie, N Mibu, S Miyano, Y Nakashima, K Watanabe, T Yamaguchi
ABSTRACT
Two pyrazine derivatives [fructosazine (3) and deoxyfructosazine (6)] were simultaneously formed in a solution of D-glucosamine hydrochloride under various conditions. They showed deoxyribonucleic acid (DNA) strand breakage activity in plasmid pBR322 comparable to that of D-glucosamine. The DNA strand breakage by fructosazine (3) was stimulated by Cu2+.