MilliporeSigma
Pseudolaric acids: isolation, bioactivity and synthetic studies.
Natural product reports (2010-04-21)
Pauline Chiu, Lai To Leung, Ben C B Ko
ABSTRACT
The pseudolaric acids are diterpenoids isolated from the root bark of Pseudolarix amabilis, or the golden larch. Pseudolaric acids A and B are the major antifungal and anti-angiogenic congeners of this family of compounds. This review presents the results of the isolation, biological and synthetic studies of these natural products. 127 references are cited.