MilliporeSigma
  • Cytokine release and cytotoxicity in human keratinocytes induced by polycyclic aromatic hydrocarbons (1-methylpyrene and perylene).
Cytokine release and cytotoxicity in human keratinocytes induced by polycyclic aromatic hydrocarbons (1-methylpyrene and perylene).
Journal of toxicology and environmental health. Part A (2010-04-15)
Raouf Bahri, Dalila Saidane-Mosbahi, Mahmoud Rouabhia
ABSTRACT
The cytotoxic effects of two polycyclic aromatic hydrocarbons (PAH) (1-methyplyrene and perylene) were investigated on human skin keratinocytes. Normal human keratinocytes were cultured in the presence of various concentrations of 1-methylpyrene and perylene either alone or in combination. Following incubation, keratinocyte adhesion, viability, proliferation, colony-forming efficiency, and apoptosis/necrosis level were examined. The effects of PAH on wound healing were also determined in vitro using a scrape-wound healing assay on epidermis-like tissue. In addition, the inflammatory cell response to PAH insult was examined through interleukin-1 (IL-1) alpha and interleukin-6 (IL-6) secretion. Each individual PAH significantly decreased keratinocyte adhesion and viability in a concentration-dependent manner, which was associated with a reduced ability of keratinocytes to proliferate and form colonies. When PAH were combined, a greater effect on keratinocyte adhesion, viability, and proliferation was noted. Decreased cell proliferation/colony-forming efficiency was accompanied by increased cell apoptosis following incubation with either PAH. This effect was enhanced by the inhibitory influence on keratinocyte migration, as assessed by culture scratching. Each PAH also exerted a significant effect on keratinocyte immune functions by modulating the secretion of inflammatory mediators. Indeed, 1-methylpyrene or perylene, individually or when combined, significantly upregulated IL-1alpha and IL-6 secretion. This effect was greater and was concentration dependent when the PAH combination was used. Overall results indicate that 1-methylpyrene and perylene exerted a cytotoxic effect on human keratinocytes. Our findings may shed light on mechanisms underlying potential adverse effects of 1-methylpyrene and perylene on human skin.
Sigma-Aldrich
1-Methylpyrene, suitable for fluorescence, ≥97.0% (GC)
Pricing and availability is not currently available.