MilliporeSigma
  • Severe central nervous system and cardiovascular toxicity in a pediatric patient after ingestion of an over-the-counter local anesthetic.
Severe central nervous system and cardiovascular toxicity in a pediatric patient after ingestion of an over-the-counter local anesthetic.
Pediatric emergency care (2009-10-17)
Jamie Nelsen, Michael Holland, Michelle Dougherty, Jason Bernad, Christine Stork, Jeanna Marraffa
ABSTRACT
Dibucaine is considered one of the most potent and consequently toxic amide anesthetics available, and despite withdrawal from the US market as a spinal anesthetic, it remains accessible as an over-the-counter preparation in the United States. Dibucaine exposures in children are infrequently encountered, but to date, all reported consequential ingestions have resulted in death. We report the first case of a potentially fatal dibucaine-induced wide-complex arrhythmia in a child who survived her clinical course without sequelae. It is our hope that this report will highlight the toxicity of dibucaine and prompt a review of its over-the-counter status. The rationale and success of a new antidote, 20% lipid emulsion, for the management of local anesthetic toxicity is discussed.