MilliporeSigma
  • Impact of metal coordination on cytotoxicity of 3-aminopyridine-2-carboxaldehyde thiosemicarbazone (triapine) and novel insights into terminal dimethylation.
Impact of metal coordination on cytotoxicity of 3-aminopyridine-2-carboxaldehyde thiosemicarbazone (triapine) and novel insights into terminal dimethylation.
Journal of medicinal chemistry (2009-07-30)
Christian R Kowol, Robert Trondl, Petra Heffeter, Vladimir B Arion, Michael A Jakupec, Alexander Roller, Markus Galanski, Walter Berger, Bernhard K Keppler
ABSTRACT
The first metal complexes of 3-aminopyridine-2-carboxaldehyde thiosemicarbazone (Triapine) were synthesized. Triapine was prepared by a novel three-step procedure in 64% overall yield. In addition, a series of related ligands, namely, 2-formylpyridine thiosemicarbazone, 2-acetylpyridine thiosemicarbazone, 2-pyridineformamide thiosemicarbazone, and their N(4)-dimethylated derivatives (including the N(4)-dimethylated analogue of Triapine) were prepared, along with their corresponding gallium(III) and iron(III) complexes with the general formula [M(L)(2)](+), where HL is the respective thiosemicarbazone. The compounds were characterized by elemental analysis, (1)H and (13)C NMR, IR and UV-vis spectroscopies, mass spectrometry, and cyclic voltammetry. In addition, Triapine and its iron(III) and gallium(III) complexes were studied by X-ray crystallography. All ligands and complexes were tested for their in vitro antiproliferative activity in two human cancer cell lines (41M and SK-BR-3), and structure-activity relationships were established. In general, the coordination to gallium(III) increased the cytotoxicity while the iron(III) complexes show reduced cytotoxic activity compared to the metal-free thiosemicarbazones. Selected compounds were investigated for the capacity of inhibiting ribonucleotide reductase by incorporation of (3)H-cytidine into DNA.
Sigma-Aldrich
3-AP, ≥98% (HPLC)