MilliporeSigma
  • One-pot multicomponent synthesis of 1-aryl-5-methyl-N-R2-1H-1,2,3-triazole-4-carboxamides: an easy procedure for combinatorial chemistry.
One-pot multicomponent synthesis of 1-aryl-5-methyl-N-R2-1H-1,2,3-triazole-4-carboxamides: an easy procedure for combinatorial chemistry.
Journal of combinatorial chemistry (2009-04-23)
Nazariy T Pokhodylo, Vasyl S Matiychuk, Mykola D Obushak
ABSTRACT
A convenient synthetic protocol was elaborated for creation of combinatorial libraries of 1-(R(1)-phenyl)-5-methyl-N-R(2)-1H-1,2,3-triazole-4-carboxamides. As starting materials, commercially available or readily prepared azides, amines, and diketene were selected for the reaction which has proceeded in a one-pot system with high yields and in short time.