MilliporeSigma
Combined MBR with photocatalysis/ozonation for bromoamine acid removal.
Applied biochemistry and biotechnology (2009-01-14)
Yuan-yuan Qu, Qing Yang, Ji-ti Zhou, Min Gou, Lin-lin Xing, Fang Ma
ABSTRACT
A combined biological (augmented membrane bioreactor (MBR)) and photochemical (photocatalysis and ozonation) treatment has been proposed for bromoamine acid (BAA) removal in dyeing wastewater. It was demonstrated that the color and chemical oxygen demand removal in the sequential augmented MBR was about 90% and 50%, respectively. By ribosomal intergenic spacer analysis, it was found that the introduced strain QYY was maintained as the predominant species and the diversity of the system was relatively low throughout the operation. Photocatalysis and ozonation processes were efficient to treat the effluents from MBR with high color and total organic carbon removal more than 90% within 120 min. Therefore, the hybrid treatment system is a possible way to achieve the complete mineralization of BAA.
Sigma-Aldrich
Bromaminic acid sodium salt
Pricing and availability is not currently available.