MilliporeSigma
Iminosugars from Baphia nitida Lodd.
Phytochemistry (2008-01-15)
Atsushi Kato, Noriko Kato, Saori Miyauchi, Yuka Minoshima, Isao Adachi, Kyoko Ikeda, Naoki Asano, Alison A Watson, Robert J Nash
ABSTRACT
Chromatographic separation of the 50% aqueous EtOH extract of the leaves of the African medicinal tree Baphia nitida resulted in isolation of 10 iminosugars. The plant contained 2R,5R-dihydroxymethyl-3R,4R-dihydroxypyrrolidine (DMDP) as a major alkaloid. The structure of a new alkaloid was also elucidated by spectroscopic methods as the 1-O-beta-D-fructofuranoside of DMDP, and this plant produced 3-O-beta-D-glucopyranosyl-DMDP as well. DMDP is a potent inhibitor of beta-glucosidase and beta-galactosidase, whereas the other two derivatives lowered inhibition toward both of these enzymes and improved inhibitory activities toward rice alpha-glucosidase and rat intestinal maltase.