MilliporeSigma
  • Gas chromatographic analysis of isomeric organic mononitrates in plasma.
Gas chromatographic analysis of isomeric organic mononitrates in plasma.
Journal of chromatography (1991-11-15)
T B Tzeng, H L Fung
ABSTRACT
A specific, sensitive and precise capillary gas chromatographic method using electron-capture detection was developed for the determination of four isomeric vasodilating organic mononitrates, viz. L-isoidide mononitrate (L-IIMN), isosorbide-2-mononitrate (IS-2-MN), isomannide mononitrate (IMMN) and isosorbide-5-mononitrate (IS-5-MN), in rat plasma. With a sample size of 100 microliters of rat plasma, the detection limits were found to be between 0.5 and 2 ng/ml for these mononitrates, and the absolute recovery was found to range from 83 to 90%. The within-day coefficients of variation for the assay of the four isomers were less than 5%, while the between-day coefficients of variation were less than 10%. Because of the short retention times of these isomers in this assay, routine analyses of about sixty plasma samples per day can be carried out. The possibility of in vivo interconversion among these four isomers in rats was investigated after individual administration of each isomer. No interconversion was found based on examination of plasma samples. The gas chromatographic method was applied to the pharmacokinetic studies of these four isomers in rats; at an intravenous dose of 2 mg/kg, the biological half-lives of L-IIMN, IMMN, IS-2-MN and IS-5-MN were found to be 13.2, 25.2, 54.6 and 112 min, respectively.
Product Description
Isosorbide 2-nitrate, European Pharmacopoeia (EP) Reference Standard