MilliporeSigma
  • Using 'cling film' to protect lower limb plaster casts in babies with club foot.
Using 'cling film' to protect lower limb plaster casts in babies with club foot.
British journal of nursing (Mark Allen Publishing) (2007-12-13)
Nirav K Patel, P J S Jeer, Mark S Cornell
ABSTRACT
Clubfoot or congenital talipes equinovarus is a common condition affecting babies and non-surgical treatment involves serial manipulation and plaster casting for many weeks. The casts are susceptible to soiling during this time, which makes management and child care even more challenging. The authors report initial experience in a typical district general hospital clubfoot clinic where the parents of a baby used conventional cling film to cover the casts and provide a low-cost, effective and well-tolerated method of protection. Informal reports received from these parents were very positive and encouraging throughout the duration of treatment. The authors believe parents with babies undergoing such treatment for clubfoot could be advised of the benefits of using cling film to protect plaster casts. More formal analysis of feedback from parents and collaborative experience with other hospitals is required before widespread use is recommended. There may also be scope for using cling film to protect lower limb casts used in managing developmental dysplasia of the hip or fractures in children and potentially adults.