MilliporeSigma
[Chemical constituents of Morinda officinalis How].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1991-11-01)
S Li, Q Ouyang, X Tan, S Shi, Z Yao
ABSTRACT
This paper reports the identification of four compounds isolated from the cortex of Morinda officinalis growing in Guangdong Province. These compounds are beta-sitosterol (I), 2-methyl-anthraquinone(II), rubiadin-1-methyl ether(III) and 24-ethylcholesterol(VI). Compounds II and VI are isolated from Morinda Genus for the first time.
Sigma-Aldrich
2-Methylanthraquinone, technical, ≥95% (HPLC)