MilliporeSigma
  • Characterization of human cytochrome P450 enzymes involved in the metabolism of cyamemazine.
Characterization of human cytochrome P450 enzymes involved in the metabolism of cyamemazine.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2007-10-24)
Christophe Arbus, Amine Benyamina, Pierre-Michel Llorca, Franck Baylé, Norbert Bromet, Frédéric Massiere, Ricardo P Garay, Ahcène Hameg
ABSTRACT
Recombinant human liver microsomal enzymes of the cytochrome P450 family (CYP1A2, CYP2A6, CYP3A4, CYP2B6, CYP2C8, CYP2C9, CYP2C19, CYP2D6, CYP2E1) were used to determine the metabolic fate of the antipsychotic anxiolytic agent cyamemazine. An LC/MS-MS tandem methodology was developed specifically for identifying the presence of cyamemazine and its metabolites in reaction media. All P450 enzymes investigated, with the exception of CYP2A6 and CYP2E1, degraded cyamemazine, albeit to a different extent, with CYP1A2, CYP2C8 and CYP2C19 being the most efficient (>80%). However, in microsomes prepared from native human hepatocytes, only relatively specific competitors (inhibitors and/or substrates) of CYP1A2, CYP2C8, CYP2C9 and CYP3A4 reduced notably the degradation cyamemazine. The main routes of cyamemazine biotransformation are N-mono-demethylation (CYP1A2, CYP3A4 and CYP2C8) and mono-oxidation (either S-oxidized or hydroxylated derivatives which could not be discriminated because characterized by the same mass value) by CYP1A2 and CYP2C9. Secondary metabolic routes yields N,N-di-demethylated and N-demethylated mono-oxidized products. Thus, under in vitro conditions, cyamemazine is extensively degraded by at least four distinct P450 enzymes, into two primary hydrophilic metabolites. These results suggest that cyamemazine detoxification process is unlikely to be significantly impaired by co-administration of therapeutic agents that are substrates of the CYP metabolic system.
Sigma-Aldrich
Cyamemazine, ≥98% (HPLC)