MilliporeSigma
  • Hit generation and exploration: imidazo[4,5-b]pyridine derivatives as inhibitors of Aurora kinases.
Hit generation and exploration: imidazo[4,5-b]pyridine derivatives as inhibitors of Aurora kinases.
Bioorganic & medicinal chemistry letters (2007-10-16)
Vassilios Bavetsias, Chongbo Sun, Nathalie Bouloc, Jóhannes Reynisson, Paul Workman, Spiros Linardopoulos, Edward McDonald
ABSTRACT
A hit generation and exploration approach led to the discovery of 31 (2-(4-(6-chloro-2-(4-(dimethylamino)phenyl)-3H-imidazo[4,5-b]pyridin-7-yl)piperazin-1-yl)-N-(thiazol-2-yl)acetamide), a potent, novel inhibitor of Aurora-A, Aurora-B and Aurora-C kinases with IC(50) values of 0.042, 0.198 and 0.227microM, respectively. Compound 31 inhibits cell proliferation and has good microsomal stability.
Sigma-Aldrich
4-Azabenzimidazole, 99%