MilliporeSigma
  • Deorphanization of Dresden G protein-coupled receptor for an odorant receptor.
Deorphanization of Dresden G protein-coupled receptor for an odorant receptor.
Journal of receptor and signal transduction research (2007-09-22)
Yoshifumi Fujita, Tomoko Takahashi, Akiko Suzuki, Kayo Kawashima, Futoshi Nara, Ryuta Koishi
ABSTRACT
Dresden G protein-coupled receptor (D-GPCR) is one of orphan G protein-coupled receptors (GPCR). Here we report the identification of the ligands and the characterization of D-GPCR. We investigated over 5000 compounds to evoke the response mediated by D-GPCR and identified 3-methyl-valeric acid and 4-methyl-valeric acid as agonists using a cAMP assay. It is of interest that they dramatically enhanced the intracellular cAMP accumulation and the CRE-luciferase activity in CHO-K1 cells and HEK293 cells expressing the chimeric protein of D-GPCR with a rhodopsin-tag at its N-terminus. Our results established new characteristics of D-GPCR as an olfactory receptor. First, agonists of D-GPCR belong to odorants. Second, D-GPCR mRNA is expressed in the olfactory bulb. In addition, D-GPCR was reported to have similar sequences and its genome locus nearby other olfactory receptors. These results suggest D-GPCR is an olfactory receptor.
Sigma-Aldrich
3-Methylvaleric acid, 97% (GC)
Sigma-Aldrich
3-Methylpentanoic acid, ≥98%, FG