MilliporeSigma
  • Femtosecond IR spectroscopy of peroxycarbonate photodecomposition: S1-lifetime determines decarboxylation rate.
Femtosecond IR spectroscopy of peroxycarbonate photodecomposition: S1-lifetime determines decarboxylation rate.
The journal of physical chemistry. A (2007-09-18)
Christian Reichardt, Jörg Schroeder, Dirk Schwarzer
ABSTRACT
The ultrafast photofragmentation of arylperoxycarbonates R-O-C(O)O-O-tert-butyl (R = naphthyl, phenyl) is studied using femtosecond UV excitation at 266 nm and mid-infrared broadband probe pulses to elucidate the dissociation mechanism. Our experiments show that the rate of fragmentation is determined by the S1-lifetime of the peroxide, i.e., the time constants of S1 decay and of CO2 and R-O* formation are identical. The fragmentation times are solvent dependent and for tert-butyl-2-naphthylperoxycarbonate (TBNC) vary from 25 ps in CH2Cl2 to 52 ps in n-heptane. In the case of the tert-butylphenylperoxycarbonate (TBPC) the decomposition takes 5.5 ps in CD2Cl2 and 12 ps in n-heptane. The CO2 fragment is formed vibrationally hot with an excess energy of about 5000 cm(-1). The hot CO2 spectra at high energy can be modeled assuming Boltzmann distributions with initial vibrational temperatures of ca. 2500 K which relax to ambient temperature with time constants of 280 ps in CCl4 and 130 ps in n-heptane. In CCl4 the relaxed spectra at 1.5 ns show 3.5% residual excitation in the n = 1 level of the asymmetric stretch vibration.
Product Description
Erythromycin ethyl succinate, European Pharmacopoeia (EP) Reference Standard