MilliporeSigma
  • A fluorescent fluoride ion probe based on a self-organized ensemble of 5-hydroxyflavone-Al(III) complex.
A fluorescent fluoride ion probe based on a self-organized ensemble of 5-hydroxyflavone-Al(III) complex.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-05-29)
R Sai Sathish, A Goutam Raju, G Nageswara Rao, C Janardhana
ABSTRACT
In the presence of Al(III) ions, 5-hydroxyflavone (5HF) through a complexation reaction in MeOH, shows dual fluorescence, characterized by a newly developed peak at 554nm upon excitation at 363nm. In this communication, the subsequent ligand exchange reaction of the complex with fluoride ion causing a fluorescence enhancement followed by a decrease in fluorescence intensity involving an intermediate mechanistic pathway, delivering a quantitative estimation route for fluoride ion in the concentration range from 5x10(-5) to 7x10(-4)M, has been reported. The ligand exchange reaction, without interference from other common anions, has been investigated by UV-vis and fluorescence spectroscopies combined with the AM1 semi-empirical self-consistent field quantum chemical calculations within UHF formalism in their ground state.