- Synthesis and pharmacological characterization of N3-substituted willardiine derivatives: role of the substituent at the 5-position of the uracil ring in the development of highly potent and selective GLUK5 kainate receptor antagonists.
Synthesis and pharmacological characterization of N3-substituted willardiine derivatives: role of the substituent at the 5-position of the uracil ring in the development of highly potent and selective GLUK5 kainate receptor antagonists.
Some N3-substituted analogues of willardiine such as 11 and 13 are selective kainate receptor antagonists. In an attempt to improve the potency and selectivity for kainate receptors, a range of analogues of 11 and 13 were synthesized with 5-substituents on the uracil ring. An X-ray crystal structure of the 5-methyl analogue of 13 bound to GLUK5 revealed that there was allowed volume around the 4- and 5-positions of the thiophene ring, and therefore the 4,5-dibromo and 5-phenyl (67) analogues were synthesized. Compound 67 (ACET) demonstrated low nanomolar antagonist potency on native and recombinant GLUK5-containing kainate receptors (KB values of 7 +/- 1 and 5 +/- 1 nM for antagonism of recombinant human GLUK5 and GLUK5/GLUK2, respectively) but displayed IC50 values >100 microM for antagonism of GLUA2, GLUK6, or GLUK6/GLUK2.