MilliporeSigma
  • Arylamine N-acetyltransferase responsible for acetylation of 2-aminophenols in Streptomyces griseus.
Arylamine N-acetyltransferase responsible for acetylation of 2-aminophenols in Streptomyces griseus.
Journal of bacteriology (2006-12-13)
Hirokazu Suzuki, Yasuo Ohnishi, Sueharu Horinouchi
ABSTRACT
An arylamine N-acetyltransferase (NAT) responsible for the N acetylation of exogenous 3-amino-4-hydroxybenzoic acid in Streptomyces griseus was identified and characterized. This enzyme was distinct from other eukaryotic and bacterial NATs in that it acetylated various 2-aminophenol derivatives more effectively than it acetylated 5-aminosalicylic acid, and thus it may be involved in the metabolism of xenobiotic compounds.
Sigma-Aldrich
3-Amino-4-hydroxybenzoic acid, 97%