MilliporeSigma
  • Electrochemical degradation of 4-chloroguaiacol for wastewater treatment using PbO2 anodes.
Electrochemical degradation of 4-chloroguaiacol for wastewater treatment using PbO2 anodes.
Journal of hazardous materials (2006-07-18)
Y Samet, S Chaabane Elaoud, S Ammar, R Abdelhedi
ABSTRACT
Electrochemical oxidation of 4-chloroguaiacol (4-CG) at Nb/PbO(2) anodes was studied under different experimental conditions such as initial concentration of substrate, electrolysis time, temperature and pH. We measured the concentrations of 4-chlorocatechol (4-CC), 2-methoxyhydroquinone (2-MHQ), maleic acid (MA) and carbon dioxide as the main products. Black solid particles consisting mainly of polymers were formed during electrolysis. A mechanism of electrochemical oxidation of 4-CG was investigated. The oxidation of 4-CG can generally be described by simple pseudo first-order kinetics. The degradation of 4-CG was favoured at high temperature and lower initial concentration of 4-CG and low solution pH. However, the increase of temperature has not a significant effect on the mineralization of carboxylic acids. Moreover, these products required long electrolysis time.
Sigma-Aldrich
4-Chlorocatechol, 97%