- Reflex activity caused by laryngoscopy and intubation is obtunded differently by meptazinol, nalbuphine and fentanyl.
Reflex activity caused by laryngoscopy and intubation is obtunded differently by meptazinol, nalbuphine and fentanyl.
To evaluate the different potencies of several opioids in obtunding reflex mechanisms of laryngoscopy and intubation. Three groups of patients (each n = 25, ASA 1-2) undergoing elective plastic surgery were randomly given meptazinol (2.5 mg kg-1), nalbuphine (0.3 mg kg-1) or fentanyl (5 microg kg-1) in a blinded fashion prior to laryngoscopy and intubation. This was followed by a standardized bolus induction of a barbiturate and a muscle relaxant. The response to laryngoscopy and intubation was studied, using blood pressure, heart rate and bispectral index. With fentanyl, there was an increase of heart rate by 17%, and systolic blood pressure by 7% when compared to control. Bispectral index dropped an additional 8% when compared to 1 min after barbiturate induction. In the nalbuphine group there was a 16% increase in systolic blood pressure, and a 16% increase in heart rate when compared to control. Also, bispectral index increased by 18% when compared to 1 min after barbiturate injection. The group receiving meptazinol demonstrated no cardiovascular changes although bispectral index dropped by an additional 19% when compared to 1 min after barbiturate injection. Meptazinol, appears to depress cardiovascular stimulatory effects and electroencephalogram arousal induced by laryngoscopy and intubation better than nalbuphine or fentanyl.