MilliporeSigma
  • Comparison of advanced oxidation processes and identification of monuron photodegradation products in aqueous solution.
Comparison of advanced oxidation processes and identification of monuron photodegradation products in aqueous solution.
Chemosphere (2005-11-18)
Maria Bobu, Steven Wilson, Tyge Greibrokk, Elsa Lundanes, Ilie Siminiceanu
ABSTRACT
The photodegradation of monuron (3-(4-chlorophenyl)-1,1-dimethylurea) in aqueous solutions under simulated solar irradiation has been conducted by different advanced oxidation processes (UV/H(2)O(2), UV/H(2)O(2)/Fe(2+), UV/H(2)O(2)/TiO(2), UV/TiO(2), dark H(2)O(2)/Fe(3+)). The degradation rates were always higher for the homogeneous catalysis in photo-Fenton reactions (UV/H(2)O(2)/Fe(2+)) compared to the heterogeneous photocatalytic systems (TiO(2)/UV and UV/H(2)O(2)/TiO(2)). Optimal concentrations of Fe(2+) and H(2)O(2) for the abatement of the herbicide in the photo-Fenton system were found to be 1 mM Fe(II) and 10 mM H(2)O(2). Several intermediary products were identified using large volume injection micro-liquid chromatography with UV detection (mu-LC-UV), mu-LC-MS and GC-MS techniques and a degradation mechanism has been proposed.