MilliporeSigma
  • Acute effects of cholestatic and choleretic bile salts on vasopressin- and glucagon-induced hepato-biliary calcium fluxes in the perfused rat liver.
Acute effects of cholestatic and choleretic bile salts on vasopressin- and glucagon-induced hepato-biliary calcium fluxes in the perfused rat liver.
The Biochemical journal (1992-04-15)
Y Hamada, A Karjalainen, B A Setchell, J E Millard, F L Bygrave
ABSTRACT
The effects were investigated of the choleretic bile salt glycoursodeoxycholate (G-UDCA) and of the cholestatic bile salt taurochenodeoxycholate (T-CDCA) on changes in perfusate Ca2+, glucose and oxygen and in bile calcium and bile flow induced by the administration of (a) vasopressin, (b) glucagon and (c) glucagon plus vasopressin together to the perfused rat liver [Hamada, Karjalainen, Setchell, Millard & Bygrave (1992) Biochem. J. 281, 387-392]. G-UDCA itself increased the secretion of calcium in the bile several-fold, but its principal effect was to augment each of the above-mentioned metabolic events except glucose and oxygen output; particularly noteworthy was its ability to augment the 'transients' in bile calcium and bile flow seen immediately after the administration of vasopressin with or without glucagon. T-CDCA, by contrast, produced opposite effects and attenuated all of the parameters measured, and in particular the transients in bile calcium and bile flow. The data provide evidence of a strong correlation between calcium fluxes occurring on both the sinusoidal and the bile-canalicular membranes and that all are modifiable by glucagon, Ca(2+)-mobilizing hormones and bile salts.
Sigma-Aldrich
Glycoursodeoxycholic acid, ≥96.0% (TLC)