MilliporeSigma
Beta-turns induced in bradykinin by (S)-alpha-methylproline.
FEBS letters (1992-02-10)
J H Welsh, O Zerbe, W von Philipsborn, J A Robinson
ABSTRACT
The ability of (S)-alpha-methylproline (alpha-MePro) to stabilise reverse-turn conformations in the peptide hormone bradykinin (BK = Arg1-Pro2-Pro3-Gly4-Phe5-Ser6-Pro7-Phe8-Arg9) has been investigated. Two BK analogues containing alpha-MePro at position 3 or position 7 were synthesised and their conformations in aqueous solution investigated by NMR spectroscopy. Whereas BK is largely disordered on the NMR time scale both analogues showed ROE connectivities in 2D-ROESY spectra indicative of reverse-turn conformations at both Pro2-Phe5 and Ser6-Arg9, whose formation appears to be cooperative. Some potential applications of alpha-MePro as a reverse-turn mimetic in the construction of synthetic peptide libraries is discussed.
Sigma-Aldrich
α-Methyl-L-proline, ≥98.0% (TLC)