MilliporeSigma
  • Cholinesterase inhibitory activity of two farnesylacetone derivatives from the brown alga Sargassum sagamianum.
Cholinesterase inhibitory activity of two farnesylacetone derivatives from the brown alga Sargassum sagamianum.
Archives of pharmacal research (2003-11-12)
Geonseek Ryu, Soo Hee Park, Eun Sook Kim, Byoung Wook Choi, Shi Yong Ryu, Bong Ho Lee
ABSTRACT
Two known farnesylacetone derivatives (1 and 2) were isolated from the Korean brown alga Sargassum sagamianum off Jeju Island, Korea. Compounds 1 and 2 were identified as (5E,10Z)-6,10,14-trimethylpentadeca-5,10-dien-2,12-dione and (5E,9E,13E)-6,10,4-trimethyl-pentadeca-5,9,13-trien-2,12-dione, respectively, by comparison with the literature data. Compounds 1 and 2 showed moderate acetylcholinesterase and butyrylcholinesterase inhibitory activities with IC50 values of 65.0 approximately 48.0 and 34.0 approximately 23.0 microM, respectively.