- Sublethal effects of insecticides on adult longevity and fecundity of German cockroaches (Dictyoptera: Blattellidae).
Sublethal effects of insecticides on adult longevity and fecundity of German cockroaches (Dictyoptera: Blattellidae).
The effects of sublethal concentrations of chlorpyrifos, cyfluthrin, and hydramethylnon on adult longevity and fecundity of German cockroaches, Blattella germanica (L.), were investigated. Longevity of males declined linearly with increasing doses of insecticide. An LD50 of cyfluthrin decreased male longevity by 52%, whereas an LD50 of hydramethylnon reduced male longevity by 81%. Longevity of females increased linearly with increasing sublethal doses of chlorpyrifos, whereas all doses of both cyfluthrin and hydramethylnon decreased longevity of females. Fecundity increased linearly with increasing sublethal doses of chlorpyrifos. Number of oothecae formed, oothecae hatched, and number of offspring produced in each ootheca increased with increasing sublethal concentrations of chlorpyrifos. In contrast, fecundity declined with increasing sublethal concentrations of cyfluthrin and hydramethylnon. Formation of the first ootheca occurred approximately 8 d after mating for untreated females but generally longer with sublethal concentrations of all insecticides. The period between oothecae hatch and the formation of subsequent oothecae increased with successive oothecae in all treatments.