MilliporeSigma
Nucleotide sequences of serine tRNAs from Bacillus subtilis.
Biochimica et biophysica acta (1992-04-06)
J Matsugi, H T Jia, K Murao, H Ishikura
ABSTRACT
Three B. subitilis serine tRNAs were sequenced including modified nucleosides. All the serine tRNAs contained 1-methyl-adenosine in the D-loop. As other characteristic modified nucleosides, 5-methoxyuridine was found in the first letter of the anticodon in the tRNA(UGA).