MilliporeSigma
  • Role of heme oxygenase-1 protein in the neuroprotective effects of cyclopentenone prostaglandin derivatives under oxidative stress.
Role of heme oxygenase-1 protein in the neuroprotective effects of cyclopentenone prostaglandin derivatives under oxidative stress.
The European journal of neuroscience (2003-06-20)
Takumi Satoh, Megumi Baba, Daisaku Nakatsuka, Yasuyuki Ishikawa, Hiroyuki Aburatani, Kyoji Furuta, Toshihisa Ishikawa, Hiroshi Hatanaka, Masaaki Suzuki, Yasuyoshi Watanabe
ABSTRACT
Previously we found that some cyclopenteone prostaglandin derivatives (PGs), referred to as neurite outgrowth-promoting PGs (NEPPs), have dual biological activities of promoting neurite outgrowth and preventing neuronal death [Satoh et al. (2000) J. Neurochem., 75, 1092-1102; Satoh et al. (2001) J. Neurochem., 77, 50-62; Satoh et al. (2002) In Kikuchi, II. (ed.), Strategenic Medical Science Against Brain Attack. Springer-Verlag, Tokyo, pp. 78-93]. To investigate possible cellular mechanisms of the neuroprotective effects, we performed oligo hybridization-based DNA array analysis with mRNA isolated from HT22, a cell line that originated from a mouse hippocampal neuron. Several transcripts up-regulated by NEPP11 were identified. Because heme oxygenase 1 (HO-1) mRNA was the most prominently induced and was earlier reported to protect neuronal and non-neuronal cells against oxidative stress, we focused on it as a possible candidate responsible for the neuroprotective effects. We found NEPP11 to induce HO-1 protein (32 kDa) in HT22 cells in both the presence and the absence of glutamate, whereas non-neuroprotective prostaglandins (PGs) Delta12-PGJ2 or PGA2 did not. Overexpression of HO-1-green fluorescence protein (GFP) fusion protein significantly protected HT22 cells against oxidative glutamate toxicity, whereas that of GFP alone did not. Furthermore, biliverdin and bilirubin, products of HO-1 enzymatic activity on heme, protected HT22 cells from oxidative glutamate toxicity. These results, together with our previous results, suggest that NEPP11 activates the expression of HO-1 and that HO-1 produces biliverdin and bilirubin, which result in the inhibition of neuronal death induced by oxidative stress. NEPP11 is the first molecular probe reported to have a neuroprotective action through induction of HO-1 in neuronal cells.