MilliporeSigma
  • Development and validation of analytical methods for monomeric and oligomeric migrants from nylon 12 packaging materials.
Development and validation of analytical methods for monomeric and oligomeric migrants from nylon 12 packaging materials.
Food additives and contaminants (2003-05-31)
N H Stoffers, F Brandl, J P H Linssen, R Franz
ABSTRACT
Analytical methods for the determination of laurolactam--the monomer of nylon 12--as well as the cyclic dimer and trimer were established. High performance liquid chromatography using ultraviolet (HPLC-UV) and mass spectrometric detection (HPLC-MS) were both found suitable to identify and quantify monomer, cyclic dimer and trimer well below the specific migration limit (SML) of laurolactam, being 5 mg/kg of food (simulant). Gas chromatography with flame ionisation detection (GC-FID) showed to be an appropriate method for the detection of only laurolactam in aqueous and fatty food simulants. Food simulants could be analysed directly by all three methods, or after a change of solvents. For olive oil, a method for sample clean-up by size-exclusion chromatography (SEC) was established.
Sigma-Aldrich
Nylon 12, pellets